siRNA Details
| Gene Symbol: Asxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270988.1 | NP_001257917.1 | 9, 10 | 1352 | ||
| NM_172421.5 | NP_766009.2 | 8, 9 | 1208 | |||
| GenBank mRNA | AK016141.1 | - | 8, 9 | 914 | ||
| AK035679.1 | - | 3, 4 | 1780 | |||
| AK036839.1 | - | 8, 9 | 1165 | |||
| AK080679.1 | - | 9, 10 | 1326 | |||
| AK148015.1 | - | 8, 9 | 1213 | |||
| BC141140.1 | - | 9, 10 | 1186 | 9055752 | ||

Asxl2
Genomic Map