siRNA Details
| Gene Symbol: Ccdc134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172428.2 | NP_766016.2 | 7 | 1533 | ||
| GenBank mRNA | AK009756.1 | - | 1 | 687 | ||
| AK028514.1 | - | 5 | 4411 | |||
| AK132115.1 | - | 6 | 1348 | |||
| AK145747.1 | - | 7 | 1481 | |||
| AK154557.1 | - | 7 | 1561 | |||
| AK207532.1 | - | 1 | 170 | |||
| AK210740.1 | - | 1 | 170 | |||
| AK212584.1 | - | 1 | 170 | |||
| AK213819.1 | - | 1 | 170 | |||
| AK216239.1 | - | 1 | 168 | |||
| BC072586.1 | - | 7 | 1477 | 6828468 | ||

Ccdc134
Genomic Map