siRNA Details
| Gene Symbol: Celf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289613.1 | NP_001276542.1 | 13 | 2971 | ||
| NM_001289616.1 | NP_001276545.1 | 14 | 2369 | |||
| NM_001289620.1 | NP_001276549.1 | 13 | 2312 | |||
| NM_172434.3 | NP_766022.1 | 13 | 2884 | |||
| GenBank mRNA | AK019578.1 | - | 10 | 2795 | ||
| AK030259.1 | - | 12 | 2299 | |||
| AK044506.1 | - | 14 | 2333 | |||
| AK087536.1 | - | 13 | 2274 | |||
| AY165052.1 | - | 13 | 2602 | |||
| BC031740.1 | - | 9 | 3232 | |||
| BC057553.1 | - | 13 | 3027 | 6401356 | ||

Celf3
Genomic Map