siRNA Details
| Gene Symbol: Dtx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172442.3 | NP_766030.3 | 9 | 3148 | ||
| GenBank mRNA | AK086624.1 | - | 8 | 1717 | ||
| BC044798.1 | - | 4 | 1036 | |||
| BC058647.1 | - | 9 | 2605 | 6405881 | ||
| BC069975.1 | - | 8 | 1688 | |||

Dtx4
Genomic Map