siRNA Details
| Gene Symbol: Exog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172136.1 | NP_001165607.1 | 7 | 2258 | ||
| NM_172456.3 | NP_766044.1 | 6 | 2223 | |||
| GenBank mRNA | AK028887.1 | - | 6 | 2188 | ||
| AK051300.1 | - | 7 | 2222 | |||
| AK148426.1 | - | 6 | 2204 | |||
| AK162566.1 | - | 6 | 2186 | |||
| AK163555.1 | - | 6 | 2188 | |||
| BC052171.1 | - | 5 | 2029 | |||
| BC059920.1 | - | 7 | 2201 | |||
| BC064730.1 | - | 7 | 2273 | |||
| BC094589.1 | - | 4 | 4710 | |||

Exog
Genomic Map