siRNA Details
| Gene Symbol: 4932411E22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172534.2 | NP_766122.1 | 3, 4 | 481 | ||
| GenBank mRNA | AK029964.1 | - | 3, 4 | 482 | ||
| BC140961.1 | - | 3, 4 | 467 | 9055573 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4932411E22Rik| Gene Symbol: 4932411E22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172534.2 | NP_766122.1 | 3, 4 | 481 | ||
| GenBank mRNA | AK029964.1 | - | 3, 4 | 482 | ||
| BC140961.1 | - | 3, 4 | 467 | 9055573 | ||