siRNA Details
| Gene Symbol: Tmem120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172541.2 | NP_766129.1 | 1, 2 | 125 | ||
| GenBank mRNA | AK028119.1 | - | 1, 2 | 126 | ||
| BC046757.1 | - | 1, 2 | 85 | 5693443 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem120a| Gene Symbol: Tmem120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172541.2 | NP_766129.1 | 1, 2 | 125 | ||
| GenBank mRNA | AK028119.1 | - | 1, 2 | 126 | ||
| BC046757.1 | - | 1, 2 | 85 | 5693443 | ||