siRNA Details
| Gene Symbol: Adgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127353.2 | NP_001120825.2 | 12 | 1472 | ||
| GenBank mRNA | AK078911.1 | - | 3 | 453 | ||
| AK134038.1 | - | 1 | 13 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adgb| Gene Symbol: Adgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127353.2 | NP_001120825.2 | 12 | 1472 | ||
| GenBank mRNA | AK078911.1 | - | 3 | 453 | ||
| AK134038.1 | - | 1 | 13 | |||