siRNA Details
| Gene Symbol: Unk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286006.1 | NP_001272935.1 | 2 | 194 | ||
| NM_172569.4 | NP_766157.1 | 2 | 194 | |||
| GenBank mRNA | AK046395.1 | - | 2 | 189 | ||
| AK077635.1 | - | 2 | 195 | |||
| AK078095.1 | - | 2 | 185 | |||
| AK129438.1 | - | 2 | 172 | |||
| AK134361.1 | - | 2 | 314 | |||
| BB619163.1 | - | 2 | 195 | |||
| BC039964.1 | - | 2 | 156 | |||
| BC054452.1 | - | 2 | 131 | 6491704 | ||

Unk
Genomic Map