siRNA Details
| Gene Symbol: Nhlrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172501.2 | NP_766089.1 | 3 | 381 | ||
| GenBank mRNA | AK029465.1 | - | 3 | 376 | ||
| AK033167.1 | - | 3 | 381 | |||
| AK133986.1 | - | 331 | ||||
| BC087903.1 | - | 3 | 368 | 30250042 | ||
| BC107356.1 | - | 3 | 369 | 40055875 | ||

Nhlrc3
Genomic Map