siRNA Details
| Gene Symbol: Dse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172508.2 | NP_766096.1 | 2 | 752 | ||
| GenBank mRNA | AK031737.1 | - | 2 | 439 | ||
| AK045065.1 | - | 2 | 659 | |||
| AK150737.1 | - | 2 | 916 | |||
| AK153326.1 | - | 2 | 425 | |||
| BC053095.1 | - | 2 | 752 | 6401066 | ||

Dse
Genomic Map