siRNA Details
Gene Symbol: Arhgef19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172520.2 | NP_766108.1 | 4 | 733 | ||
GenBank mRNA | AK031443.1 | - | 4 | 734 | ||
AK053075.1 | - | 4 | 734 | |||
BC060376.1 | - | 2 | 463 | 5340007 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
