siRNA Details
| Gene Symbol: Megf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134399.1 | NP_001127871.1 | 9 | 1184 | ||
| NM_172522.4 | NP_766110.3 | 8 | 1091 | |||
| GenBank mRNA | AK032661.1 | - | 8 | 903 | ||
| AK051642.1 | - | 8 | 1028 | |||
| AK122555.2 | - | 9 | 1124 | |||
| BC151157.1 | - | 9 | 1085 | 9088045 | ||
| BC172024.1 | - | 9 | 1085 | 9054310 | ||

Megf11
Genomic Map