siRNA Details
Gene Symbol: Tmem120a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172541.2 | NP_766129.1 | 2 | 184 | ||
GenBank mRNA | AK028119.1 | - | 2 | 185 | ||
BC046757.1 | - | 2 | 144 | 5693443 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem120a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172541.2 | NP_766129.1 | 2 | 184 | ||
GenBank mRNA | AK028119.1 | - | 2 | 185 | ||
BC046757.1 | - | 2 | 144 | 5693443 |