siRNA Details
| Gene Symbol: Unk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286006.1 | NP_001272935.1 | 3 | 436 | ||
| NM_172569.4 | NP_766157.1 | 3 | 436 | |||
| GenBank mRNA | AK046395.1 | - | 3 | 431 | ||
| AK077635.1 | - | 3 | 437 | |||
| AK078095.1 | - | 3 | 427 | |||
| AK129438.1 | - | 3 | 414 | |||
| AK134361.1 | - | 3 | 556 | |||
| BB619163.1 | - | 3 | 437 | |||
| BC039964.1 | - | 3 | 398 | |||
| BC054452.1 | - | 3 | 373 | 6491704 | ||

Unk
Genomic Map