siRNA Details
| Gene Symbol: Tmem63c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172583.2 | NP_766171.1 | 12 | 1086 | ||
| GenBank mRNA | AK030062.1 | - | 12 | 1078 | ||
| AK034410.1 | - | 11 | 1009 | |||
| AK043536.1 | - | 11 | 1007 | |||
| BC056936.1 | - | 12 | 1086 | 5695989 | ||
| BC057088.1 | - | 12 | 1086 | 5695989 | ||

Tmem63c
Genomic Map