siRNA Details
| Gene Symbol: Lrtm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172207.1 | NP_001165678.1 | 5 | 2102 | ||
| NM_172492.3 | NP_766080.1 | 5 | 2273 | |||
| GenBank mRNA | AK039006.1 | - | 5 | 2107 | ||
| AK045494.1 | - | 5 | 2186 | |||
| BC058944.1 | - | 4 | 2663 | 6827376 | ||

Lrtm2
Genomic Map