siRNA Details
| Gene Symbol: Zswim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172503.3 | NP_766091.2 | 14 | 4233 | ||
| GenBank mRNA | AK052168.1 | - | 14 | 4247 | ||
| AK053663.1 | - | 14 | 4247 | |||
| AK084602.1 | - | 1 | 975 | |||
| AK131119.1 | - | 9 | 3091 | |||
| BC058080.1 | - | 14 | 4233 | 6822376 | ||

Zswim4
Genomic Map