siRNA Details
| Gene Symbol: Mfsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114662.1 | NP_001108134.1 | 10 | 2166 | ||
| GenBank mRNA | AK085484.1 | - | 7 | 1582 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd4| Gene Symbol: Mfsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114662.1 | NP_001108134.1 | 10 | 2166 | ||
| GenBank mRNA | AK085484.1 | - | 7 | 1582 | ||