siRNA Details
| Gene Symbol: Abhd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172511.4 | NP_766099.3 | 5 | 1361 | ||
| GenBank mRNA | AK051923.1 | - | 1 | 4550 | ||
| AK052670.1 | - | 5 | 1310 | |||
| BC058347.1 | - | 5 | 1304 | 6820515 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abhd10| Gene Symbol: Abhd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172511.4 | NP_766099.3 | 5 | 1361 | ||
| GenBank mRNA | AK051923.1 | - | 1 | 4550 | ||
| AK052670.1 | - | 5 | 1310 | |||
| BC058347.1 | - | 5 | 1304 | 6820515 | ||