siRNA Details
| Gene Symbol: Fbxo42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172518.3 | NP_766106.2 | 2 | 398 | ||
| GenBank mRNA | AK020135.1 | - | 1 | 309 | ||
| AK028867.1 | - | 2 | 244 | |||
| AK150804.1 | - | 2 | 389 | |||
| AK151503.1 | - | 2 | 355 | |||
| AK165284.1 | - | 2 | 372 | |||
| AK170202.1 | - | 2 | 400 | |||
| AK171215.1 | - | 2 | 401 | |||
| BC058667.1 | - | 2 | 389 | 6410012 | ||

Fbxo42
Genomic Map