siRNA Details
Gene Symbol: Nipal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172524.3 | NP_766112.1 | 6 | 1409 | ||
GenBank mRNA | AK035561.1 | - | 6 | 1410 | ||
BC139819.1 | - | 6 | 1400 | 40141293 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nipal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172524.3 | NP_766112.1 | 6 | 1409 | ||
GenBank mRNA | AK035561.1 | - | 6 | 1410 | ||
BC139819.1 | - | 6 | 1400 | 40141293 |