siRNA Details
| Gene Symbol: Zfp609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172536.3 | NP_766124.2 | 5 | 2911 | ||
| GenBank mRNA | AK036715.1 | - | 4 | 2644 | ||
| AK129111.1 | - | 1 | 859 | |||
| BC058943.1 | - | 4 | 2816 | |||
| BC150683.1 | - | 4 | 2620 | 9087594 | ||

Zfp609
Genomic Map