siRNA Details
| Gene Symbol: Fam117a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172543.4 | NP_766131.2 | 8 | 2079 | ||
| GenBank mRNA | AK077747.1 | - | 8 | 2080 | ||
| AK143632.1 | - | 8 | 2087 | |||
| AK167417.1 | - | 9 | 1842 | |||
| BC055690.1 | - | 8 | 2051 | 6415669 | ||

Fam117a
Genomic Map