siRNA Details
| Gene Symbol: Unk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286006.1 | NP_001272935.1 | 16 | 3622 | ||
| NM_172569.4 | NP_766157.1 | 16 | 3661 | |||
| GenBank mRNA | AK046395.1 | - | 16 | 3656 | ||
| AK077635.1 | - | 16 | 3664 | |||
| AK078095.1 | - | 16 | 3614 | |||
| AK129438.1 | - | 16 | 3639 | |||
| AK134361.1 | - | 16 | 3742 | |||
| BC003195.1 | - | 13 | 3107 | |||
| BC005545.1 | - | 3 | 1668 | |||
| BC033048.1 | - | 4 | 1861 | |||
| BC039964.1 | - | 16 | 3623 | |||
| BC054452.1 | - | 16 | 3598 | 6491704 | ||

Unk
Genomic Map