siRNA Details
| Gene Symbol: Serinc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172588.2 | NP_766176.1 | 12 | 1717 | ||
| GenBank mRNA | AK037699.1 | - | 12 | 1717 | ||
| AK136799.1 | - | 12 | 1765 | |||
| AK165334.1 | - | 12 | 1724 | |||
| BC049189.1 | - | 12 | 1717 | 6311617 | ||
| BC062131.1 | - | 12 | 1717 | 6514513 | ||

Serinc5
Genomic Map