siRNA Details
| Gene Symbol: Hectd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163471.1 | NP_001156943.1 | 2 | 298 | ||
| NM_172637.3 | NP_766225.2 | 2 | 298 | |||
| GenBank mRNA | AK029549.1 | - | 2 | 158 | ||
| AK041629.1 | - | 2 | 269 | |||
| AK043646.1 | - | 2 | 259 | |||
| AK142777.1 | - | 2 | 298 | |||
| BC137671.1 | - | 2 | 269 | 8860690 | ||
| BC144815.1 | - | 2 | 269 | 9053363 | ||

Hectd2
Genomic Map