siRNA Details
| Gene Symbol: Plekhs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164263.1 | NP_001157735.1 | 4 | 292 | ||
| NM_172641.3 | NP_766229.2 | 4 | 292 | |||
| GenBank mRNA | AK036908.1 | - | 4 | 290 | ||
| AK053921.1 | - | 4 | 290 | |||
| BC090250.1 | - | 4 | 270 | 6485933 | ||

Plekhs1
Genomic Map