siRNA Details
| Gene Symbol: Kctd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172650.2 | NP_766238.1 | 5 | 517 | ||
| GenBank mRNA | AK033491.1 | - | 5 | 518 | ||
| AK034380.1 | - | 5 | 517 | |||
| AK049362.1 | - | 5 | 515 | |||
| AK087407.1 | - | 1 | 395 | |||
| AK087865.1 | - | 5 | 518 | |||
| AK139700.1 | - | 5 | 517 | |||
| BC061458.1 | - | 5 | 505 | |||
| BC079581.1 | - | 5 | 504 | 5702630 | ||

Kctd3
Genomic Map