siRNA Details
| Gene Symbol: D2Wsu81e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172660.4 | NP_766248.3 | 3, 4 | 253 | ||
| GenBank mRNA | AK044983.1 | - | 1, 2 | 847 | ||
| AK046148.1 | - | 2, 3 | 448 | |||
| AK147162.1 | - | 3, 4 | 243 | |||
| BC078441.1 | - | 1, 2 | 400 | 5686308 | ||
| CN716107.1 | - | 3, 4 | 251 | |||

D2Wsu81e
Genomic Map