siRNA Details
| Gene Symbol: Frmd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172673.3 | NP_766261.2 | 6 | 700 | ||
| GenBank mRNA | AK030883.1 | - | 6 | 456 | ||
| AK044270.1 | - | 6 | 451 | |||
| AK048865.1 | - | 6 | 701 | |||
| AK054484.1 | - | 6 | 657 | |||
| AK146383.1 | - | 6 | 668 | |||
| BC038623.1 | - | 6 | 437 | |||
| BC057549.1 | - | 3 | 229 | 6416914 | ||
| BC062889.1 | - | 6 | 657 | 5698263 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008762214, XM_008762211, XM_008762213, XM_008762212, XM_008762215.

Frmd5
Genomic Map