siRNA Details
| Gene Symbol: Fam160a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172682.3 | NP_766270.2 | 12 | 4095 | ||
| GenBank mRNA | AK036881.1 | - | 9 | 3081 | ||
| AK076386.1 | - | 4 | 2950 | |||
| BC043298.1 | - | 10 | 3139 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam160a1| Gene Symbol: Fam160a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172682.3 | NP_766270.2 | 12 | 4095 | ||
| GenBank mRNA | AK036881.1 | - | 9 | 3081 | ||
| AK076386.1 | - | 4 | 2950 | |||
| BC043298.1 | - | 10 | 3139 | |||