siRNA Details
| Gene Symbol: Fam214b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253353.1 | NP_001240282.1 | 3 | 951 | ||
| NM_001253354.1 | NP_001240283.1 | 4 | 728 | |||
| NM_172691.3 | NP_766279.1 | 3 | 732 | |||
| GenBank mRNA | AK045811.1 | - | 3 | 716 | ||
| AK149610.1 | - | 3 | 809 | |||
| AK154748.1 | - | 2 | 1067 | |||
| AK161380.1 | - | 3 | 659 | |||
| BC044832.1 | - | 2 | 892 | |||
| BC046771.1 | - | 4 | 718 | 5703628 | ||
| BC054736.1 | - | 3 | 732 | |||
| BC071191.1 | - | 1 | 420 | 6467750 | ||

Fam214b
Genomic Map