siRNA Details
| Gene Symbol: Gba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172692.3 | NP_766280.2 | 1 | 660 | ||
| GenBank mRNA | AK089192.1 | - | 1 | 346 | ||
| AK158174.1 | - | 1 | 660 | |||
| AK173213.1 | - | 1 | 646 | |||
| BC031397.1 | - | 1 | 333 | |||
| BC056935.1 | - | 1 | 183 | 5703478 | ||

Gba2
Genomic Map