siRNA Details
| Gene Symbol: March6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172606.2 | NP_766194.2 | 7 | 813 | ||
| GenBank mRNA | AK089827.1 | - | 7 | 815 | ||
| AK132461.1 | - | 7 | 785 | |||
| CF182201.1 | - | 4 | 301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

March6| Gene Symbol: March6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172606.2 | NP_766194.2 | 7 | 813 | ||
| GenBank mRNA | AK089827.1 | - | 7 | 815 | ||
| AK132461.1 | - | 7 | 785 | |||
| CF182201.1 | - | 4 | 301 | |||