siRNA Details
| Gene Symbol: Zfp523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172617.3 | NP_766205.1 | 6, 7 | 768 | ||
| GenBank mRNA | AK028222.1 | - | 6, 7 | 769 | ||
| BC025615.1 | - | 3, 4 | 333 | |||
| BC058346.1 | - | 4, 5 | 500 | 6832233 | ||
| BC060611.1 | - | 5, 6 | 732 | 6416823 | ||

Zfp523
Genomic Map