siRNA Details
| Gene Symbol: Clic5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172621.2 | NP_766209.1 | 5 | 809 | ||
| GenBank mRNA | AK017800.1 | - | 5 | 770 | ||
| AK046522.1 | - | 5 | 798 | |||
| AK156849.1 | - | 5 | 650 | |||
| BC064037.1 | - | 5 | 794 | 1227040 | ||

Clic5
Genomic Map