siRNA Details
| Gene Symbol: F11r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172647.2 | NP_766235.1 | 2 | 169 | ||
| GenBank mRNA | AK033574.1 | - | 2 | 170 | ||
| BC021876.1 | - | 2 | 112 | 3985305 | ||
| CT010347.1 | - | 2 | 89 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F11r| Gene Symbol: F11r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172647.2 | NP_766235.1 | 2 | 169 | ||
| GenBank mRNA | AK033574.1 | - | 2 | 170 | ||
| BC021876.1 | - | 2 | 112 | 3985305 | ||
| CT010347.1 | - | 2 | 89 | |||