siRNA Details
Gene Symbol: Kctd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172650.2 | NP_766238.1 | 5 | 524 | ||
GenBank mRNA | AK033491.1 | - | 5 | 525 | ||
AK034380.1 | - | 5 | 524 | |||
AK049362.1 | - | 5 | 522 | |||
AK087407.1 | - | 1 | 402 | |||
AK087865.1 | - | 5 | 525 | |||
AK139700.1 | - | 5 | 524 | |||
BC061458.1 | - | 5 | 512 | |||
BC079581.1 | - | 5 | 511 | 5702630 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
