siRNA Details
| Gene Symbol: Tmem194b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142647.1 | NP_001136119.1 | 3 | 397 | ||
| GenBank mRNA | AK030361.1 | - | 3 | 367 | ||
| AK036693.1 | - | 3 | 357 | |||
| AK048484.1 | - | 3 | 354 | |||
| CD553439.1 | - | 3 | 325 | |||

Tmem194b
Genomic Map