siRNA Details
| Gene Symbol: Tlk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172664.3 | NP_766252.2 | 8 | 862 | ||
| GenBank mRNA | AK029773.1 | - | 7 | 588 | ||
| AK172897.1 | - | 2 | 126 | |||
| BC033931.1 | - | 6 | 424 | |||
| BC051641.1 | - | 8 | 862 | 6507968 | ||
| BC057369.1 | - | 9 | 848 | |||

Tlk1
Genomic Map