siRNA Details
| Gene Symbol: Ambra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080754.1 | NP_001074223.1 | 2, 3 | 498 | ||
| NM_172669.3 | NP_766257.3 | 2, 3 | 498 | |||
| GenBank mRNA | AK037516.1 | - | 2, 3 | 455 | ||
| AK083596.1 | - | 2, 3 | 455 | |||
| AK084557.1 | - | 2, 3 | 479 | |||
| AK148887.1 | - | 2, 3 | 490 | |||
| AK155513.1 | - | 2, 3 | 461 | |||
| AK169280.1 | - | 2, 3 | 451 | |||
| BC032293.1 | - | 2, 3 | 421 | |||
| BC054082.1 | - | 2, 3 | 430 | |||

Ambra1
Genomic Map