siRNA Details
| Gene Symbol: Foxj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290696.1 | NP_001277625.1 | 4, 5 | 639 | ||
| NM_172699.3 | NP_766287.1 | 4, 5 | 639 | |||
| GenBank mRNA | AK082840.1 | - | 4, 5 | 640 | ||
| AK129271.1 | - | 4, 5 | 612 | |||
| AK133640.1 | - | 4, 5 | 609 | |||
| AK139508.1 | - | 4, 5 | 573 | |||
| BC058231.1 | - | 4, 5 | 612 | 6314981 | ||
| CB245622.1 | - | 2, 3 | 320 | |||
| CJ048244.1 | - | 4, 5 | 609 | |||

Foxj3
Genomic Map