siRNA Details
| Gene Symbol: Dpp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172624.3 | NP_766212.2 | 12 | 1549 | ||
| GenBank mRNA | AK039652.1 | - | 11 | 1642 | ||
| AK050021.1 | - | 12 | 1491 | |||
| AK050023.1 | - | 12 | 1551 | |||
| AK078301.1 | - | 12 | 1497 | |||
| AK131178.1 | - | 9 | 3412 | |||
| AK201854.1 | - | 1 | 54 | |||
| AK204395.1 | - | 1 | 54 | |||
| BC057631.1 | - | 12 | 1548 | 4191886 | ||

Dpp9
Genomic Map