siRNA Details
| Gene Symbol: Pggt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172627.3 | NP_766215.1 | 9 | 1216 | ||
| GenBank mRNA | AK028188.1 | - | 9 | 1075 | ||
| AK081731.1 | - | 9 | 1217 | |||
| BC048404.1 | - | 4 | 563 | |||
| BC141021.1 | - | 9 | 1214 | 9055633 | ||

Pggt1b
Genomic Map