siRNA Details
| Gene Symbol: Plekhs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164263.1 | NP_001157735.1 | 13 | 1651 | ||
| NM_172641.3 | NP_766229.2 | 13 | 1651 | |||
| GenBank mRNA | AK036908.1 | - | 13 | 1649 | ||
| AK053921.1 | - | 13 | 1649 | |||
| BC023093.1 | - | 3 | 1393 | |||
| BC023098.1 | - | 6 | 1002 | |||
| BC039882.1 | - | 6 | 942 | |||
| BC051054.1 | - | 3 | 1916 | |||
| BC090250.1 | - | 12 | 2644 | 6485933 | ||

Plekhs1
Genomic Map