siRNA Details
| Gene Symbol: F11r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172647.2 | NP_766235.1 | 10 | 2014 | ||
| GenBank mRNA | AK033574.1 | - | 11 | 1985 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F11r| Gene Symbol: F11r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172647.2 | NP_766235.1 | 10 | 2014 | ||
| GenBank mRNA | AK033574.1 | - | 11 | 1985 | ||