siRNA Details
Gene Symbol: Ifi205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172648.3 | NP_766236.2 | 7 | 1339 | ||
GenBank mRNA | AK040191.1 | - | 7 | 1269 | ||
BC040425.1 | - | 7 | 1339 | 3466347 | ||
CT010211.1 | - | 6 | 1187 | |||
M74124.1 | - | 6 | 1120 |
Gene Symbol: Mnda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033450.4 | NP_001028622.1 | 7 | 1503 | ||
NM_001301745.1 | NP_001288674.1 | 7 | 1497 | |||
GenBank mRNA | AK152278.1 | - | 7 | 1371 | ||
AK152683.1 | - | 7 | 1371 | |||
AK152786.1 | - | 7 | 1380 | |||
AK166217.1 | - | 7 | 1503 | |||
BC132314.1 | - | 7 | 1324 | 40130591 | ||
BC132316.1 | - | 7 | 1324 | 40130593 | ||
BC145397.1 | - | 7 | 1318 | 9053955 | ||
M74123.1 | - | 7 | 1358 | |||
S62227.1 | - | 7 | 1358 |
