siRNA Details
| Gene Symbol: Tmem194b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142647.1 | NP_001136119.1 | 9 | 1342 | ||
| GenBank mRNA | AK030361.1 | - | 8 | 1197 | ||
| AK036693.1 | - | 9 | 1256 | |||
| AK048484.1 | - | 9 | 1253 | |||
| AK086801.1 | - | 5 | 739 | |||
| AK136177.1 | - | 5 | 830 | |||
| AK170577.1 | - | 4 | 2646 | |||
| BC028796.1 | - | 6 | 768 | |||
| BC094370.1 | - | 4 | 2286 | |||

Tmem194b
Genomic Map