siRNA Details
| Gene Symbol: D2Wsu81e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172660.4 | NP_766248.3 | 7, 8 | 687 | ||
| GenBank mRNA | AK036945.1 | - | 5, 6 | 981 | ||
| AK044983.1 | - | 5, 6 | 1281 | |||
| AK046148.1 | - | 6, 7 | 882 | |||
| AK147162.1 | - | 7, 8 | 677 | |||
| BC078441.1 | - | 6 | 845 | 5686308 | ||

D2Wsu81e
Genomic Map